Assay Details
Target Gene Details
Entrez Gene ID: | 66118 |
Gene Name: | SAP domain containing ribonucleoprotein |
Gene Aliases: |
1110005A23Rik, Cip29 |
Location: |
Chr.10:128821766-128877638 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sarnp | NM_025364.2 | NP_079640.1 | ||
| AK003453.1 | BAB22799.1 | |||
| AK018773.1 | BAB31400.1 | |||
| AK088266.1 | BAC40246.1 | |||
| AK146248.1 | BAE27010.1 | |||
| AK212934.1 | ||||
| BC027510.1 | AAH27510.1 | |||
| BC157909.1 | ||||
| BC158023.1 | ||||
| BC172063.1 |

Genomic Map