Assay Details
Target Gene Details
Entrez Gene ID: | 20393 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
Sgk |
Location: |
Chr.10:21882184-21999903 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgk1 | NM_001161845.2 | NP_001155317.2 | ||
| NM_001161847.2 | NP_001155319.1 | |||
| NM_001161848.2 | NP_001155320.1 | |||
| NM_001161849.2 | NP_001155321.1 | |||
| NM_001161850.2 | NP_001155322.1 | |||
| NM_011361.3 | NP_035491.1 | |||
| AF139638.1 | AAD43302.1 | |||
| AF205855.1 | AAF19429.1 | |||
| AK086892.1 | ||||
| AK132234.1 | BAE21048.1 | |||
| AK146037.1 | BAE26849.1 | |||
| AK146062.1 | BAE26871.1 | |||
| AK159131.1 | BAE34843.1 | |||
| AK167364.1 | BAE39461.1 | |||
| BC005720.1 | AAH05720.1 | |||
| BC070401.1 | AAH70401.1 | |||
| BY736002.1 | ||||
| BY777230.1 | ||||
| BY786122.1 | ||||
| CK626189.1 | ||||
| CN709375.1 |
More Information
Set Membership: |
|

Genomic Map