Assay Details
Target Gene Details
Entrez Gene ID: | 319880 |
Gene Name: | transmembrane and coiled coil domains 3 |
Gene Aliases: |
AW488095, C630016B22Rik, C88213, Tmcc1 |
Location: |
Chr.10:94311912-94590954 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmcc3 | NM_172051.3 | 1 | 143 | NP_742048.2 |
| AK079611.1 | BAC37701.1 | |||
| AK083129.1 | BAC38774.1 | |||
| AK132545.1 | 1 | 121 | BAE21229.1 | |
| BB619673.1 | 1 | 144 | ||
| BC095951.1 | 1 | 77 | AAH95951.1 |

Genomic Map