Assay Details
Target Gene Details
Entrez Gene ID: | 67332 |
Gene Name: | small nuclear ribonucleoprotein D3 |
Gene Aliases: |
1700043E15Rik, 2310009E13Rik, AW046420, SMD3 |
Location: |
Chr.10:75518042-75535440 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snrpd3 | NM_026095.4 | NP_080371.1 | ||
| AK005341.1 | BAB23963.1 | |||
| AK006692.1 | BAB24705.1 | |||
| AK019078.1 | ||||
| AK160321.1 | BAE35738.1 | |||
| BC011510.1 | AAH11510.1 |
More Information
Set Membership: |
|

Genomic Map