Assay Details
Target Gene Details
Entrez Gene ID: | 19386 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
A430087B05Rik, AI256741, NUP358 |
Location: |
Chr.10:58446852-58494155 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ranbp2 | NM_011240.3 | NP_035370.2 | ||
| AK040322.1 | ||||
| AK041932.1 | BAC31101.1 | |||
| AK145306.1 |
More Information
Set Membership: |
|

Genomic Map