Assay Details
Target Gene Details
Entrez Gene ID: | 70097 |
Gene Name: | SAM and SH3 domain containing 1 |
Gene Aliases: |
1100001C18Rik, 2500002E12Rik, A330076K04Rik, mKIAA0790 |
Location: |
Chr.10:8722217-8956137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sash1 | NM_175155.4 | NP_780364.3 | ||
| AJ507736.1 | CAD47812.1 | |||
| AK129216.1 | BAC98026.1 | |||
| AK149872.1 |
More Information
Set Membership: |
|

Genomic Map