Assay Details
Target Gene Details
Entrez Gene ID: | 20610 |
Gene Name: | SMT3 suppressor of mif two 3 homolog 3 (yeast) |
Gene Aliases: |
2810014B19Rik, D10Ertd345e, SMT3A, SUMO-3, Smt3h1 |
Location: |
Chr.10:77606097-77618331 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sumo3 | NM_001301671.1 | NP_001288600.1 | ||
| NM_001301672.1 | NP_001288601.1 | |||
| NM_001301673.1 | NP_001288602.1 | |||
| NM_019929.4 | NP_064313.1 | |||
| NR_125904.1 | ||||
| AF063847.1 | AAC99333.1 | |||
| AK012742.1 | BAB28442.1 | |||
| AK013019.1 | BAB28601.1 | |||
| AK148306.1 | BAE28469.1 | |||
| AK150063.1 | BAE29276.1 | |||
| AK150200.1 | BAE29374.1 | |||
| AK160169.1 | BAE35670.1 | |||
| AK162035.1 | BAE36692.1 | |||
| AK167207.1 | BAE39334.1 | |||
| AK168988.1 | BAE40788.1 | |||
| AK170896.1 | BAE42100.1 | |||
| AK171169.1 | BAE42290.1 | |||
| AK190631.1 | ||||
| BC115488.1 | ||||
| BC115489.1 | ||||
| BY085566.1 | ||||
| CJ069985.1 | ||||
| CJ143013.1 | ||||
| CJ159246.1 | ||||
| CJ187500.1 | ||||
| CK619326.1 |

Genomic Map