Assay Details
Target Gene Details
Entrez Gene ID: | 17931 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 12A |
Gene Aliases: |
1200015F06Rik, 5730577I22Rik, AA792106, AV099298, D10Ertd625e, Mypt1 |
Location: |
Chr.10:108162065-108279646 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r12a | NM_027892.2 | 21 | 2724 | NP_082168.1 |
| AK004785.1 | 21 | 2724 | BAB23563.1 | |
| AK030460.1 | 11 | 1155 | ||
| AK035230.1 | 13 | 1511 | BAC28990.1 | |
| AK162199.1 | 10 | 1156 | BAE36787.1 | |
| BC125381.1 | 22 | 2793 | ||
| BC137630.1 | 22 | 2793 |
More Information
Set Membership: |
|

Genomic Map