Assay Details
Target Gene Details
Entrez Gene ID: | 52551 |
Gene Name: | small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha |
Gene Aliases: |
5330427H01Rik, AI194281, D10Ertd190e, Sgt, Stg |
Location: |
Chr.10:81044071-81060232 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgta | NM_024499.1 | NP_078775.1 | ||
| AK039966.1 | BAC30486.1 | |||
| AK051005.1 | BAC34494.1 | |||
| AK079168.1 | BAC37566.1 | |||
| AK165557.1 | BAE38254.1 | |||
| AK201798.1 | ||||
| BC003836.1 | AAH03836.1 |
More Information
Set Membership: |
|

Genomic Map