Assay Details
Target Gene Details
Entrez Gene ID: | 100113398 |
Gene Name: | adenosine deaminase, tRNA-specific 3 |
Gene Aliases: |
A430024H01Rik |
Location: |
Chr.10:80602880-80607654 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adat3 | NM_001100606.1 | NP_001094076.1 | ||
| BY036102.1 |
Target Gene Details
Entrez Gene ID: | 56214 |
Gene Name: | secretory carrier membrane protein 4 |
Gene Aliases: |
2410022D05Rik, AI448940, AU022392, AW742369, Sc4 |
Location: |
Chr.10:80602882-80615783 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scamp4 | NM_019575.4 | NP_062521.1 | ||
| AF224721.1 | AAF35368.1 | |||
| AK019112.1 | ||||
| AK078226.1 | ||||
| AK154094.1 | BAE32371.1 | |||
| AK160755.1 | BAE35991.1 | |||
| AK168554.1 | BAE40428.1 | |||
| BC018215.1 | AAH18215.1 | |||
| BC054104.2 | AAH54104.1 |
More Information
Set Membership: |
|

Genomic Map