Assay Details
Target Gene Details
Entrez Gene ID: | 216148 |
Gene Name: | SHC (Src homology 2 domain containing) transforming protein 2 |
Gene Aliases: |
6720466E06, SCK, ShcB, Sli |
Location: |
Chr.10:79617934-79637918 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shc2 | NM_001024539.1 | NP_001019710.1 | ||
| AK032878.1 | BAC28066.1 |
More Information
Set Membership: |
|

Genomic Map