Assay Details
Target Gene Details
Entrez Gene ID: | 21665 |
Gene Name: | thymine DNA glycosylase |
Gene Aliases: |
E130317C12Rik, JZA-3, Jza1 |
Location: |
Chr.10:82629247-82650494 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdg | NM_011561.2 | NP_035691.2 | ||
| NM_172552.3 | NP_766140.2 | |||
| AF102875.1 | AAC78146.1 | |||
| AK053870.1 | BAC35566.1 | |||
| AK164090.1 | BAE37621.1 | |||
| AK164643.1 | BAE37857.1 | |||
| AK169768.1 | BAE41355.1 | |||
| BC010315.1 | AAH10315.2 | |||
| BC085470.1 | AAH85470.1 | |||
| BC085471.1 | AAH85471.1 |
More Information
Set Membership: |
|

Genomic Map