Assay Details
Target Gene Details
Entrez Gene ID: | 378431 |
Gene Name: | taxilin beta |
Gene Aliases: |
2310001N14Rik, Mdp77 |
Location: |
Chr.10:17796091-17845665 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Txlnb | NM_138628.3 | 4 | 785 | NP_619534.2 |
| AF422244.1 | 4 | 722 | AAL33909.1 | |
| AF422245.1 | 4 | 722 | AAL33910.1 | |
| AK052324.1 | 4 | 749 | BAC34937.1 | |
| AK084639.1 | 4 | 786 | BAC39238.2 | |
| AK085904.1 | 4 | 679 | ||
| AK137442.1 | 4 | 700 | BAE23352.1 | |
| BC141023.1 | 4 | 698 |
More Information
Set Membership: |
|

Genomic Map