Assay Details
Target Gene Details
Entrez Gene ID: | 64058 |
Gene Name: | PERP, TP53 apoptosis effector |
Gene Aliases: |
1110017A08Rik, KCP1, KRTCAP1, PIGPC1, THW |
Location: |
Chr.10:18845071-18857073 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Perp | NM_022032.4 | 2 | 403 | NP_071315.1 |
| AF249870.1 | 2 | 404 | AAF64306.1 | |
| AK003734.1 | 2 | 344 | BAB22968.1 | |
| AK009184.1 | 2 | 341 | BAB26126.1 | |
| AK009190.1 | 2 | 342 | BAB26130.1 | |
| BC021772.1 | 2 | 328 | AAH21772.1 |

Genomic Map