Assay Details
Target Gene Details
Entrez Gene ID: | 19075 |
Gene Name: | DNA primase, p49 subunit |
Gene Aliases: |
AI324982 |
Location: |
Chr.10:128015215-128030030 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prim1 | NM_008921.2 | NP_032947.1 | ||
| AK011456.1 | ||||
| AK088557.1 | BAC40422.1 | |||
| AK089063.1 | BAC40729.1 | |||
| AK133878.1 | BAE21907.1 | |||
| AK160831.1 | ||||
| AK169632.1 | ||||
| BC089544.1 | AAH89544.1 | |||
| D13544.1 | BAA02744.1 | |||
| J04620.1 | 1 | 46 | AAA39880.1 |
More Information
Set Membership: |
|

Genomic Map