Assay Details
Target Gene Details
Entrez Gene ID: | 278279 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 2 |
Gene Aliases: |
8430438D04Rik, D330034A10Rik |
Location: |
Chr.10:105187663-105574482 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmtc2 | NM_177368.4 | 4 | 2279 | NP_796342.2 |
| AK052344.1 | 4 | 2252 | BAC34946.1 | |
| BC092226.1 | 4 | 2235 | AAH92226.1 | |
| BC138362.1 | 4 | 2010 | ||
| BC138363.1 | 4 | 2010 |
More Information
Set Membership: |
|

Genomic Map