Assay Details
Target Gene Details
Entrez Gene ID: | 237500 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 3 |
Gene Aliases: |
9130014E20Rik, B130008E12Rik, mSmile |
Location: |
Chr.10:100443902-100487347 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmtc3 | NM_001033332.2 | NP_001028504.1 | ||
| NM_001110013.1 | NP_001103483.1 | |||
| AK044857.1 | BAC32122.1 | |||
| AK149759.1 | ||||
| AK158192.1 | ||||
| AK165838.1 | BAE38405.1 |

Genomic Map