Assay Details
Target Gene Details
Entrez Gene ID: | 70428 |
Gene Name: | polymerase (RNA) III (DNA directed) polypeptide B |
Gene Aliases: |
2700078H01Rik, A330032P03Rik, C85372, RPC2 |
Location: |
Chr.10:84621456-84727178 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr3b | NM_027423.1 | NP_081699.1 | ||
| AK039244.1 | ||||
| AK039366.1 | 1 | 39 | BAC30327.1 | |
| AK165886.1 | 1 | 105 | BAE38438.1 | |
| BC068143.1 | AAH68143.1 |
More Information
Set Membership: |
|

Genomic Map