Assay Details
Target Gene Details
Entrez Gene ID: | 192188 |
Gene Name: | stabilin 2 |
Gene Aliases: |
FEEL-2, FELL, MFEEL-2, STAB-2 |
Location: |
Chr.10:86841210-87007942 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 58 - Exon 59 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stab2 | NM_138673.2 | NP_619614.1 | ||
| AB105393.1 | ||||
| AB105398.1 | ||||
| AB105399.1 | ||||
| AF364951.2 | AAL91684.2 | |||
| AK034522.1 | BAC28741.1 | |||
| DQ786775.1 |
More Information
Set Membership: |
|

Genomic Map