Assay Details
Target Gene Details
Entrez Gene ID: | 20587 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 |
Gene Aliases: |
AU020204, Baf47, Ini1, SNF5/INI1, Snf5 |
Location: |
Chr.10:75896769-75921614 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarcb1 | NM_001161853.1 | NP_001155325.1 | ||
| NM_011418.2 | NP_035548.1 | |||
| AB017343.1 | BAB12427.1 | |||
| AB017344.1 | BAB12428.1 | |||
| AB041578.1 | BAA95062.1 | |||
| AJ011739.1 | CAA09760.1 | |||
| AJ011740.1 | CAA09761.1 | |||
| AK008558.1 | ||||
| AK075906.1 | BAC36045.1 | |||
| AK145552.1 | BAE26504.1 | |||
| AK147638.1 | ||||
| AK147685.1 | ||||
| AK150169.1 | BAE29357.1 | |||
| BC025163.1 | AAH25163.1 |
More Information
Set Membership: |
|

Genomic Map