Assay Details
Target Gene Details
Entrez Gene ID: | 192188 |
Gene Name: | stabilin 2 |
Gene Aliases: |
FEEL-2, FELL, MFEEL-2, STAB-2 |
Location: |
Chr.10:86841210-87007942 on Build GRCm38 |
Assay Gene Location: | Within Exon 66 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stab2 | NM_138673.2 | 66 | 7379 | NP_619614.1 |
| AB105393.1 | 66 | 7379 | ||
| AB105398.1 | 66 | 7775 | ||
| AB105399.1 | 65 | 7368 | ||
| AF364951.2 | 66 | 7379 | AAL91684.2 | |
| AK034522.1 | 19 | 2323 | BAC28741.1 | |
| DQ786775.1 | 66 | 7375 |
More Information
Set Membership: |
|

Genomic Map