Assay Details
Target Gene Details
Entrez Gene ID: | 21853 |
Gene Name: | timeless circadian clock 1 |
Gene Aliases: |
C77407, Debt69, tim |
Location: |
Chr.10:128232063-128252941 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timeless | NM_001136082.2 | NP_001129554.1 | ||
| NM_001164080.1 | NP_001157552.1 | |||
| NM_001164081.1 | NP_001157553.1 | |||
| NM_011589.2 | NP_035719.1 | |||
| AB015598.1 | BAA36500.1 | |||
| AB019001.2 | BAA76390.2 | |||
| AF071506.1 | AAC79687.1 | |||
| AF098161.1 | AAC80010.1 | |||
| AF126480.1 | AAD24467.1 | |||
| AK153838.1 | BAE32206.1 | |||
| AK162012.1 | ||||
| BC026526.1 | AAH26526.1 | |||
| BC058641.1 | AAH58641.1 | |||
| BC082770.1 | AAH82770.1 | |||
| CJ088470.1 |

Genomic Map