Assay Details
Target Gene Details
Entrez Gene ID: | 216151 |
Gene Name: | polymerase (RNA) mitochondrial (DNA directed) |
Gene Aliases: |
1110018N15Rik, 4932416K13 |
Location: |
Chr.10:79736125-79746589 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polrmt | NM_172551.3 | NP_766139.2 | ||
| AK003792.2 | BAB22998.2 | |||
| AK030039.1 | ||||
| AK042071.1 | BAE20618.1 | |||
| AK047545.1 | ||||
| AK053356.1 | BAC35360.1 | |||
| AK085666.1 | BAC39500.1 | |||
| AK128956.1 | ||||
| AK154181.1 | ||||
| AK154729.1 | BAE32793.1 | |||
| BC110697.1 | AAI10698.1 | |||
| BC117940.1 |
More Information
Set Membership: |
|

Genomic Map