Assay Details
Target Gene Details
Entrez Gene ID: | 237611 |
Gene Name: | SH3 and cysteine rich domain 3 |
Gene Aliases: |
9830125E18 |
Location: |
Chr.10:127501617-127508823 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stac3 | NM_177707.3 | NP_808375.1 | ||
| AK036516.1 | BAC29460.1 | |||
| BC067208.1 | AAH67208.1 |
More Information
Set Membership: |
|

Genomic Map