Assay Details
Target Gene Details
Entrez Gene ID: | 64009 |
Gene Name: | spectrin repeat containing, nuclear envelope 1 |
Gene Aliases: |
8B, A330049M09Rik, BE692247, C130039F11Rik, CPG2, Myne1, mKIAA1756 |
Location: |
Chr.10:5020192-5550829 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 82 - Intron 82 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syne1 | NM_001079686.1 | NP_001073154.1 | ||
| NM_022027.2 | NP_071310.2 | |||
| AF281869.1 | AAG24392.1 | |||
| AF281870.1 | AAG24393.1 | |||
| AK036828.1 | BAC29595.1 | |||
| BC041779.1 | AAH41779.1 | |||
| BC054456.1 | AAH54456.1 | |||
| BC110428.1 |

Genomic Map