Assay Details
Target Gene Details
Entrez Gene ID: | 19714 |
Gene Name: | REV3-like, catalytic subunit of DNA polymerase zeta RAD54 like (S. cerevisiae) |
Gene Aliases: |
Rev, Rev3, Sez4 |
Location: |
Chr.10:39732099-39875210 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 24 - Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rev3l | NM_011264.3 | NP_035394.2 | ||
| AB031049.1 | BAA90768.1 | |||
| AF083464.1 | AAC98785.1 | |||
| AK162489.1 | BAE36943.1 | |||
| D78644.1 | BAA11461.1 |
More Information
Set Membership: |
|

Genomic Map