Assay Details
Target Gene Details
Entrez Gene ID: | 68094 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 2 |
Gene Aliases: |
5930405J04Rik |
Location: |
Chr.10:128459236-128490583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarcc2 | NM_001114096.1 | NP_001107568.1 | ||
| NM_001114097.1 | NP_001107569.1 | |||
| NM_198160.2 | NP_937803.1 | |||
| AK089461.1 | ||||
| AK146971.1 | BAE27576.1 | |||
| AK170530.1 | ||||
| BC058720.1 | AAH58720.1 | |||
| BC062102.1 | AAH62102.1 |
More Information
Set Membership: |
|

Genomic Map