Assay Details
Target Gene Details
Entrez Gene ID: | 27375 |
Gene Name: | tight junction protein 3 |
Gene Aliases: |
- |
Location: |
Chr.10:81273188-81291581 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tjp3 | NM_001282095.1 | NP_001269024.1 | ||
| NM_001282096.1 | NP_001269025.1 | |||
| NM_013769.3 | NP_038797.2 | |||
| AF157006.1 | AAF24175.1 | |||
| AK029083.1 | BAC26285.1 | |||
| AK134100.1 | BAE22013.1 | |||
| AK167587.1 | ||||
| BC012518.1 | AAH12518.1 |
More Information
Set Membership: |
|

Genomic Map