Assay Details
Target Gene Details
Entrez Gene ID: | 216227 |
Gene Name: | solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8 |
Gene Aliases: |
BC042593, Vglut3 |
Location: |
Chr.10:89574020-89621249 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a8 | NM_182959.3 | NP_892004.1 | ||
| AF510321.1 | AAN74643.1 | |||
| AK082743.1 | ||||
| AK149687.1 | BAE29026.1 | |||
| BC042593.1 | AAH42593.1 |
More Information
Set Membership: |
|

Genomic Map