Assay Details
Target Gene Details
Entrez Gene ID: | 11418 |
Gene Name: | acid-sensing (proton-gated) ion channel 2 |
Gene Aliases: |
ACIC2, Accn1, BNC1, BNaC1, BNaC1a, Mdeg |
Location: |
Chr.11:80880163-81968396 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asic2 | NM_001034013.2 | NP_001029185.1 | ||
| NM_007384.3 | NP_031410.1 | |||
| AF348465.1 | AAK40101.1 | |||
| AK134778.1 | BAE22279.1 | |||
| BC138529.1 | ||||
| BC138530.1 | ||||
| U57353.1 | AAB49183.1 | |||
| Y14634.1 | CAA74978.1 |
More Information
Set Membership: |
|

Genomic Map