Assay Details
Target Gene Details
Entrez Gene ID: | 27078 |
Gene Name: | B9 protein domain 1 |
Gene Aliases: |
AW045994, B9, Eppb9 |
Location: |
Chr.11:61505168-61512929 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B9d1 | NM_013717.2 | NP_038745.1 | ||
| AB030483.1 | BAA82643.1 | |||
| AK003038.1 | BAB22525.1 | |||
| AK010355.1 | BAB26875.1 | |||
| AK131701.1 | BAE20767.1 | |||
| BC008113.1 | AAH08113.1 |
More Information
Set Membership: |
|

Genomic Map