Assay Details
Target Gene Details
Entrez Gene ID: | 70419 |
Gene Name: | RIKEN cDNA 2810408A11 gene |
Gene Aliases: |
1700019L01Rik, 4930470J18Rik |
Location: |
Chr.11:69897358-69901721 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2810408A11Rik | NM_027419.3 | 5 | 1119 | NP_081695.2 |
| AK013042.1 | 5 | 1120 | BAB28617.1 | |
| AK015537.1 | 5 | 1045 | ||
| AK051029.1 | 5 | 1207 | ||
| AK160144.1 | 5 | 1014 | ||
| AK171674.1 | 5 | 1166 | BAE42603.1 | |
| BC005538.1 | 4 | 411 | ||
| BC051546.1 | 5 | 1080 | ||
| BC069874.1 | 5 | 1119 | AAH69874.1 |
More Information
Set Membership: |
|

Genomic Map