Assay Details
Target Gene Details
Entrez Gene ID: | 11302 |
Gene Name: | apoptosis-associated tyrosine kinase |
Gene Aliases: |
AATYK, aatyk1, mKIAA0641 |
Location: |
Chr.11:120007311-120047182 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aatk | NM_001198785.1 | 2 | 309 | NP_001185714.1 |
| NM_001198787.1 | 2 | 81 | NP_001185716.1 | |
| NM_007377.4 | 2 | 592 | NP_031403.2 | |
| AB093253.2 | 2 | 592 | BAC41437.2 | |
| AB288871.1 | 2 | 211 | ||
| AF011908.1 | 2 | 48 | AAB71837.1 | |
| AK036705.1 | 2 | 83 | BAC29541.1 | |
| AK155100.1 | 2 | 83 | BAE33045.1 | |
| AY236858.1 | 2 | 87 | AAO92350.1 | |
| AY236859.1 | 2 | 114 | AAO92351.1 | |
| BC080846.1 | 2 | 163 | AAH80846.1 |
More Information
Set Membership: |
|

Genomic Map