Assay Details
Target Gene Details
Entrez Gene ID: | 80981 |
Gene Name: | ADP-ribosylation factor-like 4D |
Gene Aliases: |
1110036H21Rik, ARL6, AW456149, Arf4l, Arfl4, Arl5 |
Location: |
Chr.11:101665541-101667832 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl4d | NM_025404.3 | NP_079680.1 | ||
| AK004126.1 | BAB23183.1 | |||
| AK013320.1 | BAB28789.1 | |||
| BC016113.1 | AAH16113.1 | |||
| BQ830332.1 |
More Information
Set Membership: |
|

Genomic Map