Assay Details
Target Gene Details
Entrez Gene ID: | 11951 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9) |
Gene Aliases: |
- |
Location: |
Chr.11:96072793-96075694 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp5g1 | NM_007506.6 | NP_031532.2 | ||
| BY297329.1 |
More Information
Set Membership: |
|

Genomic Map