Assay Details
Target Gene Details
Entrez Gene ID: | 71679 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit D |
Gene Aliases: |
0610009D10Rik |
Location: |
Chr.11:115415691-115419919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp5h | NM_027862.1 | NP_082138.1 | ||
| AF354051.1 | AAL83962.1 | |||
| AK002376.1 | BAB22053.1 | |||
| AK088617.1 | BAC40456.1 | |||
| BC016547.1 | AAH16547.1 | |||
| BC081431.1 | AAH81431.1 |
More Information
Set Membership: |
|

Genomic Map