Assay Details
Target Gene Details
Entrez Gene ID: | 217305 |
Gene Name: | CD300 molecule-like family member d |
Gene Aliases: |
4732429D16Rik, CLM5, LMIR4, MAIR-IV, clm-5 |
Location: |
Chr.11:114982443-114989886 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd300ld | NM_145437.2 | 3 | 531 | NP_663412.1 |
| AB292062.1 | 3 | 455 | ||
| AK028662.1 | 3 | 543 | ||
| AK041292.1 | 3 | 567 | BAC30893.1 | |
| AY457051.1 | 3 | 455 | AAR27942.1 | |
| BC019814.1 | 3 | 531 | AAH19814.1 |
More Information
Set Membership: |
|

Genomic Map