Assay Details
Target Gene Details
Entrez Gene ID: | 104112 |
Gene Name: | ATP citrate lyase |
Gene Aliases: |
A730098H14Rik, AW538652 |
Location: |
Chr.11:100476352-100528000 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acly | NM_001199296.1 | 25 | 3088 | NP_001186225.1 |
| NM_134037.3 | 24 | 3058 | NP_598798.1 | |
| AF332051.1 | 23 | 2853 | AAK56080.1 | |
| AF332052.1 | 23 | 2853 | AAK56081.1 | |
| AK043434.1 | 8 | 984 | ||
| AK132755.1 | 25 | 2979 | BAE21336.1 | |
| AK135528.1 | 25 | 2956 | ||
| AK136812.1 | 8 | 896 | BAE23135.1 | |
| AK149660.1 | 25 | 3067 | BAE29010.1 | |
| AK169703.1 | 24 | 2906 | BAE41315.1 | |
| BC005533.1 | 2 | 141 | AAH05533.1 | |
| BC021502.1 | 11 | 1334 | AAH21502.1 | |
| BC056378.1 | 24 | 2959 | AAH56378.1 |
More Information
Set Membership: |
|

Genomic Map