Assay Details
Target Gene Details
Entrez Gene ID: | 75452 |
Gene Name: | activating signal cointegrator 1 complex subunit 2 |
Gene Aliases: |
1700011I11Rik, 2610034L15Rik, AI482016, ASC1p100, AW046480 |
Location: |
Chr.11:4637696-4685699 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ascc2 | NM_029291.1 | NP_083567.1 | ||
AK051387.1 | BAC34621.1 | |||
AK148105.1 | BAE28346.1 | |||
AK179762.1 | ||||
AK207287.1 | ||||
BC013537.1 | AAH13537.1 | |||
BC024840.1 | AAH24840.1 |
