Assay Details
Target Gene Details
Entrez Gene ID: | 11764 |
Gene Name: | adaptor protein complex AP-1, beta 1 subunit |
Gene Aliases: |
Adtb1, b2b1660Clo |
Location: |
Chr.11:4947521-5042794 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1b1 | NM_001243043.1 | 11 | 1801 | NP_001229972.1 |
| NM_001243044.1 | 11 | 1604 | NP_001229973.1 | |
| NM_007454.3 | 11 | 1604 | NP_031480.2 | |
| AK034140.1 | 11 | 1804 | BAC28603.1 | |
| AK155892.1 | 11 | 1524 | BAE33487.1 | |
| AK159273.1 | 11 | 1495 | BAE34952.1 | |
| AK160042.1 | 11 | 1495 | BAE35584.1 | |
| AK165747.1 | 11 | 1521 | ||
| AK170376.1 | 11 | 1515 | ||
| AK177753.1 | ||||
| AK193924.1 | ||||
| BC008513.1 | 11 | 1417 | AAH08513.1 | |
| BC066827.1 | 2 | 148 | AAH66827.1 | |
| Y07919.1 | 11 | 1459 | CAA69224.1 |
More Information
Set Membership: |
|

Genomic Map