Assay Details
Target Gene Details
Entrez Gene ID: | 246782 |
Gene Name: | ATP synthase mitochondrial F1 complex assembly factor 2 |
Gene Aliases: |
ATP12, ATP12p |
Location: |
Chr.11:60400624-60418791 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atpaf2 | NM_145427.2 | NP_663402.2 | ||
| AK078500.1 | BAC37311.1 | |||
| BC013607.1 | AAH13607.1 |
More Information
Set Membership: |
|

Genomic Map