Assay Details
Target Gene Details
Entrez Gene ID: | 11764 |
Gene Name: | adaptor protein complex AP-1, beta 1 subunit |
Gene Aliases: |
Adtb1, b2b1660Clo |
Location: |
Chr.11:4947521-5042794 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1b1 | NM_001243043.1 | NP_001229972.1 | ||
| NM_001243044.1 | NP_001229973.1 | |||
| NM_007454.3 | NP_031480.2 | |||
| AK034140.1 | BAC28603.1 | |||
| AK155892.1 | BAE33487.1 | |||
| AK159273.1 | BAE34952.1 | |||
| AK160042.1 | BAE35584.1 | |||
| AK165747.1 | ||||
| AK170376.1 | ||||
| AK180383.1 | ||||
| BC008513.1 | AAH08513.1 | |||
| BY253407.1 | ||||
| Y07919.1 | CAA69224.1 |
More Information
Set Membership: |
|

Genomic Map