Assay Details
Target Gene Details
Entrez Gene ID: | 11764 |
Gene Name: | adaptor protein complex AP-1, beta 1 subunit |
Gene Aliases: |
Adtb1, b2b1660Clo |
Location: |
Chr.11:4947521-5042794 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1b1 | NM_001243043.1 | 5 | 897 | NP_001229972.1 |
| NM_001243044.1 | 5 | 700 | NP_001229973.1 | |
| NM_007454.3 | 5 | 700 | NP_031480.2 | |
| AK034140.1 | 5 | 900 | BAC28603.1 | |
| AK155892.1 | 5 | 620 | BAE33487.1 | |
| AK159273.1 | 5 | 591 | BAE34952.1 | |
| AK160042.1 | 5 | 591 | BAE35584.1 | |
| AK165747.1 | 5 | 616 | ||
| AK170376.1 | 5 | 609 | ||
| AK180593.1 | ||||
| AK212012.1 | 1 | 127 | ||
| BC008513.1 | 5 | 513 | AAH08513.1 | |
| Y07919.1 | 5 | 555 | CAA69224.1 |
More Information
Set Membership: |
|

Genomic Map