Assay Details
Target Gene Details
Entrez Gene ID: | 13139 |
Gene Name: | diacylglycerol kinase, alpha |
Gene Aliases: |
80kDa, AW146112, Dagk1 |
Location: |
Chr.10:128720134-128748420 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgka |
Target Gene Details
Entrez Gene ID: | 78428 |
Gene Name: | within bgcn homolog (Drosophila) |
Gene Aliases: |
A030010B05Rik, Pym |
Location: |
Chr.10:128747879-128766568 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wibg | NM_001253705.1 | NP_001240634.1 | ||
| NM_030100.4 | NP_084376.2 | |||
| AJ459407.1 | CAD30678.1 | |||
| AK020702.1 |

Genomic Map