Assay Details
Target Gene Details
Entrez Gene ID: | 432530 |
Gene Name: | adenylate cyclase 1 |
Gene Aliases: |
AC1, D11Bwg1392e, I-AC, brl, mKIAA4070 |
Location: |
Chr.11:7063489-7178506 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcy1 | NM_009622.1 | 9 | 1814 | NP_033752.1 |
| AF053980.1 | 9 | 1407 | AAC29478.1 | |
| AK220469.1 | 3 | 285 | BAD90496.1 |

Genomic Map