Assay Details
Target Gene Details
Entrez Gene ID: | 217030 |
Gene Name: | synergin, gamma |
Gene Aliases: |
Ap1gbp1, SYNG |
Location: |
Chr.11:83964380-84044578 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Synrg | NM_194341.2 | 15 | 2878 | NP_919322.1 |
| AK142858.1 | 2 | 876 | BAE25212.1 | |
| BC056370.1 | 15 | 2838 | AAH56370.1 |

Genomic Map