Assay Details
Target Gene Details
Entrez Gene ID: | 192197 |
Gene Name: | breast carcinoma amplified sequence 3 |
Gene Aliases: |
1500019F07Rik, 2610028P08Rik, AU021018, BC028339, K20D4, rudhira |
Location: |
Chr.11:85353116-85826058 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcas3 | NM_001166642.1 | NP_001160114.1 | ||
| NM_001166643.1 | NP_001160115.1 | |||
| NM_138681.4 | NP_619622.3 | |||
| AK032423.1 | BAC27862.1 | |||
| AK157222.1 | ||||
| AK171964.1 | BAE42749.1 | |||
| BC118965.2 | ||||
| BC118966.1 |

Genomic Map