Assay Details
Target Gene Details
Entrez Gene ID: | 14025 |
Gene Name: | B cell CLL/lymphoma 11A (zinc finger protein) |
Gene Aliases: |
2810047E18Rik, BCL-11A, Ctip1, D930021L15Rik, Evi9, Evi9a, Evi9b, Evi9c, mKIAA1809 |
Location: |
Chr.11:24076564-24173558 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl11a | NM_001159289.1 | NP_001152761.1 | ||
| NM_001159290.1 | NP_001152762.1 | |||
| NM_001242934.1 | 4 | 971 | NP_001229863.1 | |
| NM_016707.3 | 4 | 971 | NP_057916.1 | |
| AF051525.1 | 4 | 909 | AAF22430.1 | |
| AF169036.1 | AAF65928.1 | |||
| AF169037.1 | AAF65929.1 | |||
| AF186018.1 | 4 | 852 | AAF63682.1 | |
| AK004395.1 | BAB23285.1 | |||
| AK043677.1 | BAC31616.1 | |||
| AK045556.1 | BAC32416.1 | |||
| AK140949.1 | 4 | 916 | BAE24527.1 | |
| BC010585.1 | AAH10585.1 | |||
| BC051418.1 | AAH51418.1 |
More Information
Set Membership: |
|

Genomic Map