Assay Details
Target Gene Details
Entrez Gene ID: | 71770 |
Gene Name: | adaptor-related protein complex 2, beta 1 subunit |
Gene Aliases: |
1300012O03Rik, AI788979 |
Location: |
Chr.11:83298969-83405035 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap2b1 | NM_001035854.2 | NP_001030931.1 | ||
| NM_027915.3 | NP_082191.1 | |||
| AK004975.1 | BAB23711.1 | |||
| AK030505.1 | ||||
| AK032617.1 | ||||
| AK138434.1 | BAE23661.1 | |||
| BC046772.1 | AAH46772.1 |
More Information
Set Membership: |
|

Genomic Map