Assay Details
Target Gene Details
Entrez Gene ID: | 12330 |
Gene Name: | calnexin |
Gene Aliases: |
1110069N15Rik, AI988026, Cnx, D11Ertd153e |
Location: |
Chr.11:50293957-50325673 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Canx | NM_001110499.1 | 10 | 1252 | NP_001103969.1 |
| NM_001110500.1 | 10 | 1220 | NP_001103970.1 | |
| NM_007597.3 | 10 | 1256 | NP_031623.1 | |
| AK028279.1 | 10 | 1184 | ||
| AK084175.1 | 10 | 1196 | BAC39133.1 | |
| AK159298.1 | 10 | 1240 | BAE34971.1 | |
| AK165508.1 | 10 | 1155 | BAE38226.1 | |
| AK166649.1 | 10 | 1233 | BAE38916.1 | |
| BC012408.1 | 10 | 1174 | AAH12408.1 | |
| BC040244.1 | 10 | 1183 | AAH40244.1 | |
| L18888.1 | 9 | 1068 | AAA21014.1 | |
| L23865.1 | 9 | 1008 | AAA62450.1 |
More Information
Set Membership: |
|

Genomic Map